Tisk…

2-Chloro-6-(2,2,3,3-tetrafluoropropoxy)pyrazine 95%

BLDPBD01068266-250EA 3650 CZK
BLDPBD01068266-250 BLDPBD01068266-1G BLDPBD01068266-5G
2-Chloro-6-(2,2,3,3-tetrafluoropropoxy)pyrazine 95%
2-Chloro-6-(2,2,3,3-tetrafluoropropoxy)pyrazine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₅ClF₄N₂O
MW: 244,5700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11197521
CAS číslo: 1039858-65-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR