Tisk…

tert-Butyl 4-hydroxy-3,3-dimethylpiperidine-1-carboxylate 95%

BLDPBD01068800-5GEA 8300 CZK
BLDPBD01068800-5G BLDPBD01068800-1G BLDPBD01068800-250
tert-Butyl 4-hydroxy-3,3-dimethylpiperidine-1-carboxylate 95%
tert-Butyl 4-hydroxy-3,3-dimethylpiperidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₂H₂₃NO₃
MW: 229,3159 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977343
CAS číslo: 143306-65-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR