Tisk…

2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine 98%

BLDPBD01069578-100EA 656 CZK
BLDPBD01069578-100 BLDPBD01069578-5G BLDPBD01069578-1G BLDPBD01069578-250
2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine 98%
2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₃₆H₄₄N₂O₄
MW: 568,76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28386094
CAS číslo: 358727-55-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR