Tisk…

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride 95%

BLDPBD0107-5GEA 428 CZK
BLDPBD0107-5G BLDPBD0107-10G
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride 95%
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₀H₁₂ClNO₂
MW: 213,6600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800357
CAS číslo: 77497-95-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR