Tisk…

(E)-4-Fluorobenzaldehyde oxime 97%

BLDPBD01072722-25GEA 7160 CZK
BLDPBD01072722-25G BLDPBD01072722-1G BLDPBD01072722-5G
(E)-4-Fluorobenzaldehyde oxime 97%
(E)-4-Fluorobenzaldehyde oxime

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₆FNO
MW: 139,1271 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00133101
CAS číslo: 7304-35-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR