Tisk…

3,4-Dimethoxybenzene-1-sulphonyl fluoride 95%

BLDPBD01074673-100EA 1230 CZK
BLDPBD01074673-100 BLDPBD01074673-1G BLDPBD01074673-5G BLDPBD01074673-250
3,4-Dimethoxybenzene-1-sulphonyl fluoride 95%
3,4-Dimethoxybenzene-1-sulphonyl fluoride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₈H₉FO₄S
MW: 220,2181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21886431
CAS číslo: 95546-50-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR