Tisk…

Ethyl 2-hydroxy-3-phenylpropanoate 98%

BLDPBD01075021-100EA 4510 CZK
BLDPBD01075021-100 BLDPBD01075021-250 BLDPBD01075021-1G
Ethyl 2-hydroxy-3-phenylpropanoate 98%
Ethyl 2-hydroxy-3-phenylpropanoate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₁H₁₄O₃
MW: 194,2271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128177
CAS číslo: 15399-05-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR