Tisk…

4-(Fluorosulphonyl)-3-methylbenzoic acid 95%

BLDPBD01076030-100EA 4050 CZK
BLDPBD01076030-100 BLDPBD01076030-1G BLDPBD01076030-250
4-(Fluorosulphonyl)-3-methylbenzoic acid 95%
4-(Fluorosulphonyl)-3-methylbenzoic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₇FO₄S
MW: 218,2022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24488048
CAS číslo: 33866-06-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR