Tisk…

1-Chloropentan-2-one 95%

BLDPBD01081753-1GEA 9350 CZK
BLDPBD01081753-1G BLDPBD01081753-100 BLDPBD01081753-5G BLDPBD01081753-250
1-Chloropentan-2-one 95%
1-Chloropentan-2-one

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₅H₉ClO
MW: 120,5774 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16167605
CAS číslo: 19265-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR