Tisk…

6-Bromo-8-chloroisoquinoline 95%

BLDPBD01083223-250EA 6420 CZK
BLDPBD01083223-250 BLDPBD01083223-1G BLDPBD01083223-100
6-Bromo-8-chloroisoquinoline 95%
6-Bromo-8-chloroisoquinoline

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₅BrClN
MW: 242,4997 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28129772
CAS číslo: 1824270-14-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR