Tisk…

3-Ethynyl-6-methyl-pyridazine 95%

BLDPBD01084482-100EA 8240 CZK
BLDPBD01084482-100 BLDPBD01084482-1G BLDPBD01084482-250 BLDPBD01084482-25M
3-Ethynyl-6-methyl-pyridazine 95%
3-Ethynyl-6-methyl-pyridazine

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₆N₂
MW: 118,1359 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22371717
CAS číslo: 77778-20-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR