Tisk…

(((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl) methyl) ((2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl) dihydrogen diphosphate 98+%

BLDPBD01085129-250EA 12460 CZK
BLDPBD01085129-250 BLDPBD01085129-50M BLDPBD01085129-100 BLDPBD01085129-1G
(((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl) methyl) ((2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl) dihydrogen diphosphate 98+%
(((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl) methyl) ((2R,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl) dihydrogen diphosphate

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₁₆H₂₅N₅O₁₅P₂
MW: 589,3417 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00870977
CAS číslo: 15839-70-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR