Tisk…

3-Methylpentane-1,5-diyl diacrylate 95%

BLDPBD01085660-100EA 4910 CZK
BLDPBD01085660-100 BLDPBD01085660-1G BLDPBD01085660-250
3-Methylpentane-1,5-diyl diacrylate 95%
3-Methylpentane-1,5-diyl diacrylate

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₈O₄
MW: 226,2689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28968982
CAS číslo: 64194-22-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR