Tisk…

4,4'-(Anthracene-9,10-diyl)dibenzoic acid 98%

BLDPBD01089754-1GEA 3420 CZK
BLDPBD01089754-1G BLDPBD01089754-250 BLDPBD01089754-5G
4,4'-(Anthracene-9,10-diyl)dibenzoic acid 98%
4,4'-(Anthracene-9,10-diyl)dibenzoic acid

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₂₈H₁₈O₄
MW: 418,4401 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07644571
CAS číslo: 42824-53-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR