Tisk…

2,5-Bis(2-ethylhexyl)-3,6-di(furan-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 97%

BLDPBD01090089-1GEA 6160 CZK
BLDPBD01090089-1G BLDPBD01090089-250 BLDPBD01090089-100
2,5-Bis(2-ethylhexyl)-3,6-di(furan-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione 97%
2,5-Bis(2-ethylhexyl)-3,6-di(furan-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₃₀H₄₀N₂O₄
MW: 492,6500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31631763
CAS číslo: 1286755-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR