Tisk…

3'-Iodo-2'-methylacetophenone 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD01115661-1GEA 8360 CZK
BLDPBD01115661-1G BLDPBD01115661-100 BLDPBD01115661-250
3'-Iodo-2'-methylacetophenone 97%
3'-Iodo-2'-methylacetophenone

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Vzorec: C₉H₉IO
MW: 260,072 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22573017
CAS číslo: 52164-39-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR