Tisk…

2-Bromo-4-fluoro-6-nitrotoluene 98+%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD01374653-1GEA 371 CZK
BLDPBD01374653-1G BLDPBD01374653-100 BLDPBD01374653-5G BLDPBD01374653-25G
2-Bromo-4-fluoro-6-nitrotoluene 98+%
2-Bromo-4-fluoro-6-nitrotoluene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₅BrFNO₂
MW: 234,0225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094189
CAS číslo: 502496-33-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR