Tisk…

1-Bromo-4-ethoxy-2-methylbenzene 99+%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD01382840-1GEA 1370 CZK
BLDPBD01382840-1G BLDPBD01382840-25G BLDPBD01382840-100 BLDPBD01382840-5G
1-Bromo-4-ethoxy-2-methylbenzene 99+%
1-Bromo-4-ethoxy-2-methylbenzene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₉H₁₁BrO
MW: 215,087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917736
CAS číslo: 68155-69-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR