Tisk…

Geranyl chloride 95%

BLDPBD01409701-250EA 399 CZK
BLDPBD01409701-250 BLDPBD01409701-5G BLDPBD01409701-25G BLDPBD01409701-1G
Geranyl chloride 95%
Geranyl chloride

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₀H₁₇Cl
MW: 172,6950 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000983
CAS číslo: 5389-87-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR