2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 99+%
|
|
BLDPBD01422337-500EA
3990
CZK
BLDPBD01422337-500
BLDPBD01422337-25G
BLDPBD01422337-0.0
BLDPBD01422337-100
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 99+%
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₁₂H₁₆O₄ MW: 224,2530 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085267 CAS číslo: 106797-53-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...