Tisk…

6-Bromo-2,3-difluorotoluene 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD01435850-25GEA 6420 CZK
BLDPBD01435850-25G BLDPBD01435850-1G BLDPBD01435850-5G
6-Bromo-2,3-difluorotoluene 95%
6-Bromo-2,3-difluorotoluene

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₇H₅BrF₂
MW: 207,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07777161
CAS číslo: 847502-81-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR