Tisk…

2'-Amino-4',5'-dimethoxyacetophenone 96%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD0209-25GEA 6020 CZK
BLDPBD0209-25G BLDPBD0209-5G BLDPBD0209-0.025G BLDPBD0209-250MG BLDPBD0209-1G
2'-Amino-4',5'-dimethoxyacetophenone 96%
2'-Amino-4',5'-dimethoxyacetophenone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₃NO₃
MW: 195,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016646
CAS číslo: 4101-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR