(R)-3-(Boc-amino)piperidine 97%
|
|
BLDPBD0330-100GEA
4020
CZK
BLDPBD0330-100G
BLDPBD0330-25G
BLDPBD0330-5G
BLDPBD0330-500G
BLDPBD0330-10G
(R)-3-(Boc-amino)piperidine 97%
(R)-3-(Boc-amino)piperidine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₁₀H₂₀N₂O₂ MW: 200,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093382 CAS číslo: 309956-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...