Tisk…

3-Phenoxythiophene 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD0423-250MGEA 884 CZK
BLDPBD0423-250MG BLDPBD0423-5G BLDPBD0423-1G
3-Phenoxythiophene 98%
3-Phenoxythiophene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₁₀H₈OS
MW: 176,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00178747
CAS číslo: 63285-84-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR