Tisk…

2-Amino-4-(4-bromophenyl)-5-methylthiazole 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD0428-5GEA 5960 CZK
BLDPBD0428-5G BLDPBD0428-250MG BLDPBD0428-25G BLDPBD0428-1G
2-Amino-4-(4-bromophenyl)-5-methylthiazole 98%
2-Amino-4-(4-bromophenyl)-5-methylthiazole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Vzorec: C₁₀H₉BrN₂S
MW: 269,16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00170268
CAS číslo: 65705-44-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR