Tisk…

2',4'-Dihydroxy-3'-propylacetophenone 95%

BLDPBD0603-25GEA 7190 CZK
BLDPBD0603-25G BLDPBD0603-5G BLDPBD0603-1G
2',4'-Dihydroxy-3'-propylacetophenone 95%
2',4'-Dihydroxy-3'-propylacetophenone

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₁H₁₄O₃
MW: 194,2300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00068616
CAS číslo: 40786-69-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR