Tisk…

6-Ethyl-2-naphthalenol 98%

BLDPBD0785-100MGEA 1170 CZK
BLDPBD0785-100MG BLDPBD0785-1G BLDPBD0785-250MG BLDPBD0785-5G
6-Ethyl-2-naphthalenol 98%
6-Ethyl-2-naphthalenol

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₁₂H₁₂O
MW: 172,223 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368645
CAS číslo: 1999-64-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR