Tisk…

4'-Morpholinoacetophenone 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD0855-5GEA 941 CZK
BLDPBD0855-5G BLDPBD0855-100G BLDPBD0855-25G BLDPBD0855-1G
4'-Morpholinoacetophenone 98%
4'-Morpholinoacetophenone

Storage condition: Sealed in dry, Room Temperature

Vzorec: C₁₂H₁₅NO₂
MW: 205,2530 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006168
CAS číslo: 39910-98-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR