Tisk…

4-Sulphobenzoic acid monopotassium salt 98%

BLDPBD100239-100GEA 2430 CZK
BLDPBD100239-100G BLDPBD100239-5G BLDPBD100239-25G
4-Sulphobenzoic acid monopotassium salt 98%
4-Sulphobenzoic acid monopotassium salt

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₅KO₅S
MW: 240,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007509
CAS číslo: 5399-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR