Tisk…

4-Amino-3-methylbenzenesulphonamide 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD10050-25GEA 19840 CZK
BLDPBD10050-25G BLDPBD10050-10G BLDPBD10050-100MG BLDPBD10050-1G BLDPBD10050-250MG BLDPBD10050-5G
4-Amino-3-methylbenzenesulphonamide 97%
4-Amino-3-methylbenzenesulphonamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₇H₁₀N₂O₂S
MW: 186,2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466840
CAS číslo: 53297-70-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR