Tisk…

5-(Trifluoromethyl)pyridine-3-carboxylic acid 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD101438-250MGEA 456 CZK
BLDPBD101438-250MG BLDPBD101438-5G BLDPBD101438-1G BLDPBD101438-25G
5-(Trifluoromethyl)pyridine-3-carboxylic acid 98%
5-(Trifluoromethyl)pyridine-3-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₄F₃NO₂
MW: 191,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07774128
CAS číslo: 131747-40-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR