Tisk…

5-Bromo-2-hydroxybenzonitrile 98%

BLDPBD10254-100GEA 4760 CZK
BLDPBD10254-100G BLDPBD10254-5G BLDPBD10254-1G BLDPBD10254-10G BLDPBD10254-25G
5-Bromo-2-hydroxybenzonitrile 98%
5-Bromo-2-hydroxybenzonitrile

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₄BrNO
MW: 198,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143428
CAS číslo: 40530-18-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR