Tisk…

5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside 98%

BLDPBD102927-50MGEA 2740 CZK
BLDPBD102927-50MG BLDPBD102927-250MG
5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside 98%
5-Bromo-4-chloro-3-indolyl α-D-mannopyranoside

Storage condition: Sealed in dry, Store in freezer, under -20°C

Vzorec: C₁₄H₁₅BrClNO₆
MW: 408,629 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078951
CAS číslo: 125229-64-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR