Tisk…

5-Bromo-6-methyl-1H-indazole 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD103007-1GEA 599 CZK
BLDPBD103007-1G BLDPBD103007-10G BLDPBD103007-250MG BLDPBD103007-25G BLDPBD103007-100MG BLDPBD103007-5G
5-Bromo-6-methyl-1H-indazole 97%
5-Bromo-6-methyl-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₈H₇BrN₂
MW: 211,0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026991
CAS číslo: 885223-72-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR