Tisk…

3-Fluoro-5-(trifluoromethyl)phenylboronic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD10320-1GEA 770 CZK
BLDPBD10320-1G BLDPBD10320-10G BLDPBD10320-5G
3-Fluoro-5-(trifluoromethyl)phenylboronic acid 97%
3-Fluoro-5-(trifluoromethyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₅BF₄O₂
MW: 207,92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06657735
CAS číslo: 159020-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR