Tisk…

2-Carboxyphenylboronic acid 96%

BLDPBD10486-10GEA 2030 CZK
BLDPBD10486-10G BLDPBD10486-100G BLDPBD10486-5G BLDPBD10486-25G BLDPBD10486-1G
2-Carboxyphenylboronic acid 96%
2-Carboxyphenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₇H₇BO₄
MW: 165,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318118
CAS číslo: 149105-19-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR