Methyl-3-methylthiophene-2-carboxylate 98%
|
|
BLDPBD10576-25GEA
3940
CZK
BLDPBD10576-25G
BLDPBD10576-100G
BLDPBD10576-1G
BLDPBD10576-5G
BLDPBD10576-10G
Methyl-3-methylthiophene-2-carboxylate 98%
Methyl-3-methylthiophene-2-carboxylate
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Vzorec:
C₇H₈O₂S MW: 156,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234332 CAS číslo: 81452-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...