Tisk…

5-(3-(Trifluoromethyl)phenyl)oxazole 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD107142-1GEA 3940 CZK
BLDPBD107142-1G BLDPBD107142-250MG BLDPBD107142-100MG
5-(3-(Trifluoromethyl)phenyl)oxazole 95%
5-(3-(Trifluoromethyl)phenyl)oxazole

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₁₀H₆F₃NO
MW: 213,1600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00085180
CAS číslo: 175205-48-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR