Tisk…

6-Bromo-2,3-difluorotoluene 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD108352-250MGEA 656 CZK
BLDPBD108352-250MG BLDPBD108352-1G
6-Bromo-2,3-difluorotoluene 98%
6-Bromo-2,3-difluorotoluene

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₇H₅BrF₂
MW: 207,02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07777161
CAS číslo: 847502-81-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR