Tisk…

2-Bromo-2',6'-difluoroacetophenone 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD10860-1GEA 371 CZK
BLDPBD10860-1G BLDPBD10860-250MG
2-Bromo-2',6'-difluoroacetophenone 97%
2-Bromo-2',6'-difluoroacetophenone
Vzorec: C₈H₅BrF₂O
MW: 235,0255 g/mol
MDL Number: MFCD04038255
CAS číslo: 56159-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR