Tisk…

3-Chloro-6-(cyclopropylamino)pyridazine 98%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD108973-250MGEA 513 CZK
BLDPBD108973-250MG BLDPBD108973-1G BLDPBD108973-5G
3-Chloro-6-(cyclopropylamino)pyridazine 98%
3-Chloro-6-(cyclopropylamino)pyridazine

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₇H₈ClN₃
MW: 169,6100 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09800959
CAS číslo: 941294-45-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR