5-Hydroxyisatoic anhydride 97%
|
|
BLDPBD10916-1GEA
1320
CZK
BLDPBD10916-1G
BLDPBD10916-5G
BLDPBD10916-250MG
5-Hydroxyisatoic anhydride 97%
5-Hydroxyisatoic anhydride
Storage condition: Sealed in dry, Room Temperature
Vzorec:
C₈H₅NO₄ MW: 179,1296 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798201 CAS číslo: 195986-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...