Tisk…

5-Fluoro-2-methylbenzaldehyde 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD11053-1GEA 228 CZK
BLDPBD11053-1G BLDPBD11053-100G BLDPBD11053-10G BLDPBD11053-5G BLDPBD11053-25G
5-Fluoro-2-methylbenzaldehyde 97%
5-Fluoro-2-methylbenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₇FO
MW: 138,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02261763
CAS číslo: 22062-53-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR