Tisk…

4-Fluoroisatoic anhydride 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD110583-250MGEA 456 CZK
BLDPBD110583-250MG BLDPBD110583-1G BLDPBD110583-10G BLDPBD110583-5G BLDPBD110583-25G
4-Fluoroisatoic anhydride 97%
4-Fluoroisatoic anhydride

Storage condition: Inert atmosphere, 2-8°C

Vzorec: C₈H₄FNO₃
MW: 181,1200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03839826
CAS číslo: 321-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR