Tisk…

4-(Dimethylcarbamoyl)phenylboronic acid 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD11311-100GEA 14230 CZK
BLDPBD11311-100G BLDPBD11311-5G BLDPBD11311-250MG BLDPBD11311-25G BLDPBD11311-1G
4-(Dimethylcarbamoyl)phenylboronic acid 97%
4-(Dimethylcarbamoyl)phenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Vzorec: C₉H₁₂BNO₃
MW: 193,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258943
CAS číslo: 405520-68-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR