Tisk…

4'-Fluoro-2'-(trifluoromethyl)acetophenone 97%

BLDPBD11513-100GEA 11580 CZK
BLDPBD11513-100G BLDPBD11513-25G BLDPBD11513-500G BLDPBD11513-1G BLDPBD11513-10G BLDPBD11513-0.5G BLDPBD11513-5G
4'-Fluoro-2'-(trifluoromethyl)acetophenone 97%
4'-Fluoro-2'-(trifluoromethyl)acetophenone

Storage condition: Sealed in dry, 2-8°C

Vzorec: C₉H₆F₄O
MW: 206,14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00061251
CAS číslo: 208173-21-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR