Tisk…

(Cyanomethyl)triphenylphosphonium chloride 95%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD1160-5GEA 200 CZK
BLDPBD1160-5G BLDPBD1160-25G BLDPBD1160-500G BLDPBD1160-100G BLDPBD1160-10G
(Cyanomethyl)triphenylphosphonium chloride 95%
(Cyanomethyl)triphenylphosphonium chloride

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₂₀H₁₇ClNP
MW: 337,79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031672
CAS číslo: 4336-70-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR