Tisk…

Benzyloxyacetaldehyde diethyl acetal 98%

BLDPBD116314-1GEA 285 CZK
BLDPBD116314-1G BLDPBD116314-25G BLDPBD116314-100G BLDPBD116314-5G
Benzyloxyacetaldehyde diethyl acetal 98%
Benzyloxyacetaldehyde diethyl acetal

Storage condition: Inert atmosphere, Room Temperature

Vzorec: C₁₃H₂₀O₃
MW: 224,30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00051786
CAS číslo: 42783-78-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR