Tisk…

5-Amino-3-methyl-1-phenyl-1H-pyrazole 98%

BLDPBD11906-25GEA 228 CZK
BLDPBD11906-25G BLDPBD11906-500G BLDPBD11906-100G
5-Amino-3-methyl-1-phenyl-1H-pyrazole 98%
5-Amino-3-methyl-1-phenyl-1H-pyrazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Vzorec: C₁₀H₁₁N₃
MW: 173,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020727
CAS číslo: 1131-18-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR