Tisk…

2,4-Dimethoxy-3-methylbenzaldehyde 97%

Dodavatel: BLD PHARMATECH GMBH

BLDPBD12038-25GEA 24140 CZK
BLDPBD12038-25G BLDPBD12038-250MG BLDPBD12038-5G BLDPBD12038-1G
2,4-Dimethoxy-3-methylbenzaldehyde 97%
2,4-Dimethoxy-3-methylbenzaldehyde

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Vzorec: C₁₀H₁₂O₃
MW: 180,2005 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00011489
CAS číslo: 7149-92-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR